Learn More
Biomatik Corporation Rat Apolipoprotein C2 (APOC2) Wide-range ELISA Kit
Supplier:  Biomatik Corporation EKU10522
Encompass_Preferred
Rat Apolipoprotein C2 (APOC2) Wide-range ELISA Kit for detecting APOC2 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.69ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: G3V8D4. Gene ID: 292697. Target Synonyms: APC2; Apo-C2; ProapoC-II; Apolipoprotein C-II; Proapolipoprotein C-II. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4474