Learn More
Biomatik Corporation Rat Apolipoprotein F (APOF) ELISA Kit
Supplier:  Biomatik Corporation EKU02523
Encompass_Preferred
Rat Apolipoprotein F (APOF) ELISA Kit for detecting APOF in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.54ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q5M889. Gene ID: 500761. Target Synonyms: Apo-F; LTIP; Lipid transfer inhibitor protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6913