Learn More
American Research Products Inc Rat Apoptosis regulator BAX B
Catalog No. 5000327528
Encompass
Rat Apoptosis regulator BAX (BAX) ELISA kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27528 Supplier American Research Products Inc Supplier No. CSBEL002573RA
Rat Apoptosis regulator BAX (BAX) ELISA kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich