Learn More
Kingfisher Biotech Inc Rat APRIL (TNFSF13) Polyclonal Antibody (100 ug)
SDP
Supplier:  Kingfisher Biotech Inc PB0472R100
Encompass
The Rat APRIL (TNFSF13) polyclonal antibody is unlabeled and has been qualified for use in ELISA and Western blot applications. It is the capture antibody in the Kingfisher Biotech Mouse APRIL ELISA. The Rat APRIL polyclonal antibody was produced in goats and is antigen-affinity purified. The reactivity by species is: (Bovine APRIL - Weak) (Human APRIL - None) (Mouse APRIL - Strong) (Rat APRIL - Strong) (Swine APRIL - None). For research use only. Made in the USA
Catalog No. 50-233-2274