Learn More
American Research Products Inc Rat Arachidonate 5 lipoXygenas
Catalog No. 5000328438
Encompass
Rat Arachidonate 5-lipoXygenase (AloX5) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-28438 Supplier American Research Products Inc Supplier No. CSBE16982R
Rat Arachidonate 5-lipoXygenase (AloX5) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich