Learn More
BIOHIPPO Rat ARC Microsample
SDP
Catalog No. 5000315468
Encompass
Rat ARC Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ARC in Tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 118 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat ARC Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat ARC Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat ARC biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15468 Supplier BIOHIPPO Supplier No. BHE15210889
Rat ARC Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ARC in Tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 118 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat ARC Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat ARC Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat ARC biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color