Learn More
Cusabio Technology LLC Rat Arginase 1ARG1 ELISA Kit
Catalog No. 5000049910
Encompass_Preferred
The Rat Arginase 1 (ARG1) ELISA Kit is designed for the quantitative detection of ARG1 in serum plasma and tissue homogenates using a sandwich assay format This kit detects ARG1 within a sensitive range of 0 312 ng/mL to 20 ng/mL with a lower detection limit of 0 078 ng/mL ensuring accurate measurement of low-abundance samples The assay requires a short incubation period of 1 to 5 hours and utilizes a sample volume of 50-100 L per test Detection is performed at 450 nm following standard ELISA protocols to provide reliable and reproducible results Ideal for research applications this kit offers a robust tool for studying ARG1 expression and regulation in various biological matrices
Catalog No. 50-000-49910 Supplier Cusabio Technology LLC Supplier No. CSBE17519R
The Rat Arginase 1 (ARG1) ELISA Kit is designed for the quantitative detection of ARG1 in serum plasma and tissue homogenates using a sandwich assay format This kit detects ARG1 within a sensitive range of 0 312 ng/mL to 20 ng/mL with a lower detection limit of 0 078 ng/mL ensuring accurate measurement of low-abundance samples The assay requires a short incubation period of 1 to 5 hours and utilizes a sample volume of 50-100 L per test Detection is performed at 450 nm following standard ELISA protocols to provide reliable and reproducible results Ideal for research applications this kit offers a robust tool for studying ARG1 expression and regulation in various biological matrices