Learn More
Biomatik Corporation Rat Arginase II (Arg2) ELISA Kit
Supplier:  Biomatik Corporation EKU10662
Encompass_Preferred
Rat Arginase II (Arg2) ELISA Kit for detecting Arg2 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O08701. Gene ID: 29215. Target Synonyms: Kidney-type arginase; Non-hepatic arginase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6554