Learn More
Sino Biological Rat ART4 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 80383R08H100
Encompass_Preferred
A DNA sequence encoding the rat ART4 (NP_001166980.1)(Met1-Lys269) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-3809