Learn More
Cayman Chemical Rat Aryl Hydrocarbon RECPTR
SDP
Catalog No. 502939955
Encompass_Preferred
Rat AhR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields exceptional cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for cumbersome intermediate treatment steps such as spin-and-rinse of cells, viability determinations, cell titer adjustments, or the pre-incubation of reporter cells prior to assay setup. [INDIGO Catalog Nos. R06001-32, R06001]
Catalog No. 50-293-9955 Supplier Cayman Chemical Supplier No. 283373X32 WELL
Rat AhR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields exceptional cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for cumbersome intermediate treatment steps such as spin-and-rinse of cells, viability determinations, cell titer adjustments, or the pre-incubation of reporter cells prior to assay setup. [INDIGO Catalog Nos. R06001-32, R06001]