Learn More
American Research Products Inc Rat Aspartate Aminotransferase
Catalog No. 5000329041
Encompass
Rat Aspartate Aminotransferase (AST) ELISA kit from CUSABIO Detection Range 3 12 mIU/mL-200 mIU/mL Sensitivity 0 78 mIU/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-29041 Supplier American Research Products Inc Supplier No. CSBE13023R
Rat Aspartate Aminotransferase (AST) ELISA kit from CUSABIO Detection Range 3 12 mIU/mL-200 mIU/mL Sensitivity 0 78 mIU/mL Sample serum plasma Method Sandwich