Learn More
American Research Products Inc Rat asymmetrical dimethylargin
Catalog No. 5000326047
Encompass
Rat asymmetrical dimethylarginine (ADMA) ELISA Kit from CUSABIO Detection Range 1 5 ng/mL-300 ng/mL Sensitivity 0 75 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-26047 Supplier American Research Products Inc Supplier No. CSBE13039R
Rat asymmetrical dimethylarginine (ADMA) ELISA Kit from CUSABIO Detection Range 1 5 ng/mL-300 ng/mL Sensitivity 0 75 ng/mL Sample serum plasma tissue homogenates Method Competitive