Learn More
BIOHIPPO Rat AT Microsample
SDP
Catalog No. 5000313749
Encompass
Rat AT Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of AT in plasma It provides a detection range of 15 63-1000 ng/mL with a sensitivity of 7 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat AT Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat AT Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat AT biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13749 Supplier BIOHIPPO Supplier No. BHE15210847
Rat AT Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of AT in plasma It provides a detection range of 15 63-1000 ng/mL with a sensitivity of 7 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat AT Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat AT Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat AT biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color