Learn More
Biomatik Corporation Rat ATPase, H+ Transporting, Mitochondrial F1 Complex Beta Polypeptide (ATP5b) ELISA Kit
Supplier:  Biomatik Corporation EKU08373
Encompass_Preferred
Rat ATPase, H+ Transporting, Mitochondrial F1 Complex Beta Polypeptide (ATP5b) ELISA Kit for detecting ATP5b in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.21ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P10719. Gene ID: 171374. Target Synonyms: ATPMB; ATPSB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5361