Learn More
American Research Products Inc Rat B cell CLL lymphoma 2 BCL
Catalog No. 5000328725
Encompass
Rat B-cell CLL/lymphoma 2 (BCL2) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28725 Supplier American Research Products Inc Supplier No. CSBE08854R
Rat B-cell CLL/lymphoma 2 (BCL2) ELISA kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich