Learn More
American Research Products Inc Rat b2 microglobulin BMG b2 M
Catalog No. 5000329640
Encompass
Rat b2-microglobulin BMG/b2-MG ELISA Kit from CUSABIO Detection Range 0 156 g/mL-10 g/mL Sensitivity 0 039 g/mL Sample serum plasma urine tissue homogenates Method Sandwich
Catalog No. 50-003-29640 Supplier American Research Products Inc Supplier No. CSBE11298R
Rat b2-microglobulin BMG/b2-MG ELISA Kit from CUSABIO Detection Range 0 156 g/mL-10 g/mL Sensitivity 0 039 g/mL Sample serum plasma urine tissue homogenates Method Sandwich