Learn More
Sino Biological Recombinant Anti-Beta-2 microglobulin Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 80423R03520
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat B2M / beta-2 microglobulin (rR B2M / beta-2 microglobulin; Catalog#80423-R08H; P07151; Met1-Met119).
Catalog No. 50-240-8328