Learn More
ACROBiosystems ACROBiosystems Rat BAFFR / TNFRSF13C Protein, His Tag
Catalog No. 502106189
Encompass_Preferred
Recombinant Protein;1MG;Rat BAFFR, His Tag (BAR-R52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 10 - Asp 69 (Accession # D4A281-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 8.7 kDa. The protein migrates as 15-27 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse BAFF, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. BAF-M5257) at 5 μg/mL (100 μL/well) can bind Rat BAFFR, His Tag (Cat. No. BAR-R52H3) with a linear range of 2-78 ng/mL (QC tested).
Catalog No. 50-210-6189 Supplier ACROBiosystems Supplier No. BARR52H31MG
Recombinant Protein;1MG;Rat BAFFR, His Tag (BAR-R52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 10 - Asp 69 (Accession # D4A281-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 8.7 kDa. The protein migrates as 15-27 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse BAFF, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. BAF-M5257) at 5 μg/mL (100 μL/well) can bind Rat BAFFR, His Tag (Cat. No. BAR-R52H3) with a linear range of 2-78 ng/mL (QC tested).