Learn More
American Research Products Inc Rat basic fibroblast growth fa
Catalog No. 5000326531
Encompass
Rat basic fibroblast growth factor bFGF ELISA Kit from CUSABIO Detection Range 0 312 ng/ml -20 ng/ml Sensitivity 0 078 ng/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26531 Supplier American Research Products Inc Supplier No. CSBE08002R
Rat basic fibroblast growth factor bFGF ELISA Kit from CUSABIO Detection Range 0 312 ng/ml -20 ng/ml Sensitivity 0 078 ng/ml Sample serum plasma tissue homogenates Method Sandwich