Learn More
Sino Biological Anti-BCAM Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80222RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat BCAM (rR BCAM; Catalog#80222-R08H; Q9ESS6; Met1-Ala543). BCAM specific IgG was purified by Rat BCAM affinity chromatography.
Catalog No. 50-255-2807