Learn More
Raybiotech Inc Rat bcl-w ELISA Kit
Supplier:  Raybiotech Inc ELRBCLW1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 0.4 ng/mL. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair also can recognize human and mouse BCL-W.
Catalog No. 50-195-0216