Learn More
ACROBiosystems ACROBiosystems Rat BCMA / TNFRSF17 Protein,His Tag
Catalog No. 502107273
Encompass_Preferred
Recombinant Protein;100UG;Rat BCMA, His Tag (BCA-R52H3) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Thr 49 (Accession # D3ZKQ8-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 7.6 kDa. The protein migrates as 10-15 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BAFF, Fc Tag, active trimer (Cat. No. BAF-H5261) at 5 μg/mL (100 μL/well) can bind Rat BCMA, His Tag (Cat. No. BCA-R52H3) with a linear range of 0.039-0.625 μg/mL (QC tested).;Immobilized Rat BCMA, His Tag (Cat. No. BCA-R52H3) at 5 μg/mL (100 μL/well) can bind Anti-BCMA Neutralizing Antibody, Mouse IgG1 (Cat. No. BCA-M43) with a linear range of 0.2-5 ng/mL (Routinely tested).
Catalog No. 50-210-7273 Supplier ACROBiosystems Supplier No. BCAR52H3100UG
Recombinant Protein;100UG;Rat BCMA, His Tag (BCA-R52H3) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Thr 49 (Accession # D3ZKQ8-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 7.6 kDa. The protein migrates as 10-15 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BAFF, Fc Tag, active trimer (Cat. No. BAF-H5261) at 5 μg/mL (100 μL/well) can bind Rat BCMA, His Tag (Cat. No. BCA-R52H3) with a linear range of 0.039-0.625 μg/mL (QC tested).;Immobilized Rat BCMA, His Tag (Cat. No. BCA-R52H3) at 5 μg/mL (100 μL/well) can bind Anti-BCMA Neutralizing Antibody, Mouse IgG1 (Cat. No. BCA-M43) with a linear range of 0.2-5 ng/mL (Routinely tested).