Learn More
Sino Biological Rat Beta-2 microglobulin Matched ELISA Antibody Pair Set 15 Plates
Catalog No. 5000786180
Encompass_Preferred
The Rat Beta-2 microglobulin / B2M ELISA Pair Set is for the quantitative determination of Rat Beta-2 microglobulin / B2M.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Rat Beta-2 microglobulin / B2M coated on a 96-well plate. Standards and samples are added to the wells, and any Rat Beta-2 microglobulin / B2M present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Rat Beta-2 microglobulin / B2M monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Rat Beta-2 microglobulin / B2M present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86180 Supplier Sino Biological Supplier No. SEK8042315
The Rat Beta-2 microglobulin / B2M ELISA Pair Set is for the quantitative determination of Rat Beta-2 microglobulin / B2M.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Rat Beta-2 microglobulin / B2M coated on a 96-well plate. Standards and samples are added to the wells, and any Rat Beta-2 microglobulin / B2M present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Rat Beta-2 microglobulin / B2M monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Rat Beta-2 microglobulin / B2M present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.