Learn More
Biomatik Corporation Rat Beta-Endorphin (bEP) CLIA Kit
Supplier:  Biomatik Corporation EKU08641
Encompass_Preferred
Rat Beta-Endorphin (bEP) CLIA Kit for detecting bEP in serum, plasma, tissue homogenates, cell lysates, cerebrospinal fluid, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 7.81-2000pg/ml. Sensitivity: 3.12pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Accession Number: P01194. Target Synonyms: b-EP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5028