Learn More
Biomatik Corporation Rat Biglycan (BGN) ELISA Kit
Supplier:  Biomatik Corporation EKU10679
Encompass_Preferred
Rat Biglycan (BGN) ELISA Kit for detecting BGN in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.62ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P47853. Gene ID: 25181. Target Synonyms: DSPG1; PG-S1; PGI; SLRR1A; Biglycan Proteoglycan; Bone/cartilage proteoglycan I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8312