Learn More
BIOHIPPO Rat BMP8A Microsample
SDP
Catalog No. 5000314854
Encompass
Rat BMP8A Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of BMP8A in Serum plasma and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 107 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat BMP8A Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat BMP8A Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat BMP8A biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14854 Supplier BIOHIPPO Supplier No. BHE15210910
Rat BMP8A Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of BMP8A in Serum plasma and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 107 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat BMP8A Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat BMP8A Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat BMP8A biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color