Learn More
BIOHIPPO Rat BMPER Microsample
SDP
Catalog No. 5000314356
Encompass
Rat BMPER Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of BMPER in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 061 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat BMPER Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat BMPER Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat BMPER biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14356 Supplier BIOHIPPO Supplier No. BHE15210790
Rat BMPER Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of BMPER in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 061 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat BMPER Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat BMPER Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat BMPER biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color