Learn More
American Research Products Inc Rat bone alkaline phosphatase
Catalog No. 5000329214
Encompass
Rat bone alkaline phosphatase BALP ELISA Kit from CUSABIO Detection Range 15 6 U/L-1000 U/L Sensitivity 3 9 U/L Sample serum tissue homogenates plasma Method Sandwich
Catalog No. 50-003-29214 Supplier American Research Products Inc Supplier No. CSBE11865R
Rat bone alkaline phosphatase BALP ELISA Kit from CUSABIO Detection Range 15 6 U/L-1000 U/L Sensitivity 3 9 U/L Sample serum tissue homogenates plasma Method Sandwich