Learn More
iQ Biosciences SD Rat Bone Marrow Cells, 20M cells/vial
Catalog No. NC2120785
Encompass_Preferred
High-quality, high-viability (typically > 70%) bone marrow cells from the Sprague Dawley, rat a commonly-used outbred rat strain. Used for a wide variety of immunology-based applications, including cell population characterization and differentiation into specific cell types. All orders come with an iQ Certificate of Analysis.
This item is not returnable. View return policy
Catalog No. NC2120785 Supplier iQ Biosciences Supplier No. IQBRBM102
High-quality, high-viability (typically > 70%) bone marrow cells from the Sprague Dawley, rat a commonly-used outbred rat strain. Used for a wide variety of immunology-based applications, including cell population characterization and differentiation into specific cell types. All orders come with an iQ Certificate of Analysis.