Learn More
Biomatik Corporation Rat Bone Morphogenetic Protein 1 (BMP1) ELISA Kit
Supplier:  Biomatik Corporation EKU02739
Encompass_Preferred
Rat Bone Morphogenetic Protein 1 (BMP1) ELISA Kit for detecting BMP1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: F1M798. Gene ID: 83470. Target Synonyms: PCOLC; PCP; TLD; mTld; Procollagen C Proteinase; Mammalian tolloid protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5851