Learn More
Biomatik Corporation Rat Bone Morphogenetic Protein Receptor 2 (BMPR2) ELISA Kit
Supplier:  Biomatik Corporation EKU12109
Encompass_Preferred
Rat Bone Morphogenetic Protein Receptor 2 (BMPR2) ELISA Kit for detecting BMPR2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: F1LQC5. Target Synonyms: BMPRII; BMPR-II; BRK3; T-ALK; BMPR3; PPH1; Primary Pulmonary Hypertension 1; BMP type II receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6619