Learn More
Biomatik Corporation Rat Bone Morphogenetic Protein 9 (BMP9) CLIA Kit
Supplier:  Biomatik Corporation EKU09696
Encompass_Preferred
Rat Bone Morphogenetic Protein 9 (BMP9) CLIA Kit for detecting BMP9 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 27.4-20,000pg/ml. Sensitivity: 10.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: M0RB87. Gene ID: 290921. Target Synonyms: GDF2; Growth Differentiation Factor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8463