Learn More
American Research Products Inc Rat Bone morphogenetic protein
Catalog No. 5000326313
Encompass
Rat Bone morphogenetic protein 2 BMP-2 ELISA Kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26313 Supplier American Research Products Inc Supplier No. CSBE04508R
Rat Bone morphogenetic protein 2 BMP-2 ELISA Kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich