Learn More
American Research Products Inc Rat Bone morphogenetic protein
Supplier: American Research Products Inc CSBE04511R
Rat Bone morphogenetic protein 4 BMP-4 ELISA Kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich