Learn More
Biomatik Corporation Rat Bradykinin (BK) ELISA Kit
Supplier:  Biomatik Corporation EKU12160
Encompass_Preferred
Rat Bradykinin (BK) ELISA Kit for detecting BK in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.23-100pg/ml. Sensitivity: 0.57pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P08934. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6458