Learn More
Lonza Walkersville Rat Brain Cortex Neurons
Supplier:  Lonza Walkersville RCX500
Encompass
Ready-to-use Clonetics Rat Brain Cortex Neurons from rat (E18, 19) brain are cell suspensions of high quality primary embryonic brain neuronal cells (including glia) prepared by standardized methods, and are ready for immediate culture.Benefits:Ready-To-Use No tissue dissection or animal handling required Increased data reproducibilityCharacteristic morphology and staining tests confirm tissue origin and cell typeNegative for mycoplasma, bacteria, yeast and fungiCertificate of Analysis (CoA) is available for every batchApplications:NeurogenesisNeurotransmitter FunctionGene ExpressionSignaling PathwaysElectrophysiologyNeurotoxicityDrug/Compound ScreeningAdvanced Cell Culture Models Content:1 x Cryopreserved ampule of Rat Cortex Neurons containing ≥ 4,000,000 cells
Catalog No. NC1882237