Learn More
Lonza Walkersville Rat Brain Cx-Hi-Cp Mixed Astro
Catalog No. 5000496420
Encompass
Ready-to-use Clonetics Rat Brain Cortex Astrocytes are obtained from rat brain passaged once and prepared as cell suspensions for shipment on dry ice Glia are a mixture of rat cortical hippocampal and striatal astrocytes Each vial contains approximately 1 million cells Astrocytes can be easily thawed and cultured Astrocytes can be harvested and re-plated or stored frozen for later use The cells are shipped overnight Recommended medium AGM BulletKit (CC-3186)
Catalog No. 50-004-96420 Supplier Lonza Walkersville Supplier No. RASM530
Ready-to-use Clonetics Rat Brain Cortex Astrocytes are obtained from rat brain passaged once and prepared as cell suspensions for shipment on dry ice Glia are a mixture of rat cortical hippocampal and striatal astrocytes Each vial contains approximately 1 million cells Astrocytes can be easily thawed and cultured Astrocytes can be harvested and re-plated or stored frozen for later use The cells are shipped overnight Recommended medium AGM BulletKit (CC-3186)