Learn More
American Research Products Inc Rat Brain deriVed neurotrophic
Catalog No. 5000326301
Encompass
Rat Brain deriVed neurotrophic facor BDNF ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26301 Supplier American Research Products Inc Supplier No. CSBE04504R
Rat Brain deriVed neurotrophic facor BDNF ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich