Learn More
Lonza Walkersville Rat Brain Striatum Astrocytes
Catalog No. 5000496423
Encompass
Ready-to-use Clonetics Rat Brain Striatum Astrocytes are obtained from rat brain passaged once and prepared as cell suspensions for shipment on dry ice Each vial contains approximately 1 million cells Astrocytes can be easily thawed and cultured Astrocytes can be harvested and re-plated or stored frozen for later use The cells are shipped overnight on dry ice Recommended medium AGM BulletKit (CC-3186)
Catalog No. 50-004-96423 Supplier Lonza Walkersville Supplier No. RCPAS522
Ready-to-use Clonetics Rat Brain Striatum Astrocytes are obtained from rat brain passaged once and prepared as cell suspensions for shipment on dry ice Each vial contains approximately 1 million cells Astrocytes can be easily thawed and cultured Astrocytes can be harvested and re-plated or stored frozen for later use The cells are shipped overnight on dry ice Recommended medium AGM BulletKit (CC-3186)