Learn More
Biomatik Corporation Rat Bruton'S Tyrosine Kinase (Btk) ELISA Kit
Supplier:  Biomatik Corporation EKU02815
Encompass_Preferred
Rat Bruton'S Tyrosine Kinase (Btk) ELISA Kit for detecting Btk in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: E9PTF7. Target Synonyms: AGMX1; AT; ATK; BPK; IMD1; PSCTK1; XLA; Bruton Agammaglobulinemia Tyrosine Kinase; B-cell progenitor kinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7673