Learn More
Sino Biological Rat BST1 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 80351R08H100
Encompass_Preferred
A DNA sequence encoding the rat BST1 (Q63072) (Met1-Glu293) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-0942