Learn More
Biomatik Corporation Rat Butyrylcholinesterase (BCHE) ELISA Kit
Supplier:  Biomatik Corporation EKU10688
Encompass_Preferred
Rat Butyrylcholinesterase (BCHE) ELISA Kit for detecting BCHE in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 2.47-200ng/ml. Sensitivity: 0.95ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q9JKC1. Gene ID: 65036. Target Synonyms: BuChE; CHE1; E1; Pseudocholinesterase; Cholinesterase; Acylcholine acylhydrolase; Butyrylcholine esterase; Choline esterase II. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7504