Learn More
American Research Products Inc Rat C C chemokine receptor typ
Supplier:  American Research Products Inc CSBEL004841RA
Encompass
Rat C-C chemokine receptor type 2 (CCR2) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27769