Learn More
American Research Products Inc Rat C C motif chemokine 6 CCL
Catalog No. 5000327240
Encompass
Rat C-C motif chemokine 6 (CCL6) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-27240 Supplier American Research Products Inc Supplier No. CSBEL027415RA
Rat C-C motif chemokine 6 (CCL6) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich