Learn More
Crystal Chem Inc Rat C-Peptide ELISA Kit, 96-wells
SDP
Catalog No. 501947917
Encompass
The Rat C-Peptide ELISA is used to quantify levels of rat c-peptide in serum, plasma, and fluid using only a 5 µL sample volume. Highlights: Kits use a sample volume of only 5 µL. Dynamic Range of 0.1 - 6.4 ng/mL. Excellent cross-reactivity between C-Peptide I & II. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7917 Supplier Crystal Chem Inc Supplier No. 90055
The Rat C-Peptide ELISA is used to quantify levels of rat c-peptide in serum, plasma, and fluid using only a 5 µL sample volume. Highlights: Kits use a sample volume of only 5 µL. Dynamic Range of 0.1 - 6.4 ng/mL. Excellent cross-reactivity between C-Peptide I & II. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.