Learn More
Mercodia Inc Rat C-Peptide ELISA, Rat, Serum Plasma Cell Culture, 96 Wells
SDP
Supplier:  Mercodia Inc 10117201
Encompass_Preferred
A high quality enzyme immunoassay for the quantification of rat c-peptide in serum, plasma, or cell culture media. Measures both rat c-peptide I and II. Offers a low sample volume: 10 uL.
Catalog No. NC0540639