Learn More
Biomatik Corporation Rat C Reactive Protein (CRP) CLIA Kit
Supplier:  Biomatik Corporation EKU08919
Encompass_Preferred
Rat C Reactive Protein (CRP) CLIA Kit for detecting CRP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.14-100ng/ml. Sensitivity: 0.05ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P48199. Gene ID: 25419. Target Synonyms: C-RP; PTX1; Pentraxin-Related; Pentraxin 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5929