Learn More
Alpco Ltd Rat C-Reactive Protein ELISA
SDP
Supplier:  Alpco Ltd 41CRPRTE01
Encompass
The Rat C-Reactive Protein ELISA (Rat CRP ELISA) is intended for the quantitative measurement of IgD in rat serum and plasma. Results in less than 30 minutesRequires only 5 µL of sample. Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. NC2820437