Learn More
American Research Products Inc Rat C X C motif chemokine 9 C
Catalog No. 5000327945
Encompass
Rat C-X-C motif chemokine 9 (CXCL9) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-27945 Supplier American Research Products Inc Supplier No. CSBEL006252RA
Rat C-X-C motif chemokine 9 (CXCL9) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich