Learn More
Biomatik Corporation Rat C1q And Tumor Necrosis Factor Related Protein 3 (C1QTNF3) ELISA Kit
Supplier:  Biomatik Corporation EKU12082
Encompass_Preferred
Rat C1q And Tumor Necrosis Factor Related Protein 3 (C1QTNF3) ELISA Kit for detecting C1QTNF3 in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.65ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: B1WC91. Gene ID: 294806. Target Synonyms: CORS26; CTRP3; Corcs; Cors; Cartonectin; Collagenous repeat-containing sequence 26 kDa protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8036